6-Cyclopropyl-1H-pyrazolo[3,4-b]pyridin-3-amine ≥95%

Supplier: Matrix Scientific

048207-500MG 048207-1G
101883-298EA 538.2 USD
101883-298 101883-296
6-Cyclopropyl-1H-pyrazolo[3,4-b]pyridin-3-amine ≥95%
6-Cyclopropyl-1H-pyrazolo[3,4-b]pyridin-3-amine

Hazard: Irritant

Formula: C₉H₁₀N₄
Melting Pt: 195…197 °C
MDL Number: MFCD11841044
CAS Number: 1135283-75-8

Order Now

Specification Test Results

Melting point 195-197°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR