4-Chloro-5-methoxy-2-phenylpyrimidine ≥95%

Supplier: Matrix Scientific

048206-500MG
101883-294EA 921.54 USD
101883-294
4-Chloro-5-methoxy-2-phenylpyrimidine ≥95%
4-Chloro-5-methoxy-2-phenylpyrimidine

Hazard: Irritant

Formula: C₁₁H₉ClN₂O
Melting Pt: 155…157 °C
MDL Number: MFCD02068298
CAS Number: 13317-67-4

Order Now

Specification Test Results

Melting point 155-157°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR