Ethyl-2-(2-chloroethylidene)-1-hydrazinecarboxylate ≥95%
Supplier: Matrix Scientific
048195-5G
048195-1G
048195-500MG
101883-258EA
232.31
USD
101883-258
101883-256
101883-260
Ethyl-2-(2-chloroethylidene)-1-hydrazinecarboxylate ≥95%
Ethyl-2-(2-chloroethylidene)-1-hydrazinecarboxylate
Hazard: Irritant
Formula:
C₅H₉ClN₂O₂ Melting Pt: 120…122 °C |
MDL Number:
MFCD11227207 CAS Number: 62105-88-8 |
Specification Test Results
Melting point | 120-122°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...