4-Chloropicolinaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-2-pyridinecarbaldehyde

048191-5G 048191-1G
102663-732EA 458.46 USD
102663-732 101883-246
4-Chloropicolinaldehyde ≥97%
4-Chloropicolinaldehyde

Hazard: Irritant/Refrigerate

Formula: C₆H₄ClNO
MW: 141.5600 g/mol
Melting Pt: 32…34 °C
MDL Number: MFCD07437896
CAS Number: 63071-13-6

Order Now

Specification Test Results

Melting point 32-34°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR