1-Benzyl-3,5-dibromo-4-nitro-1H-pyrazole ≥95%
Supplier: Matrix Scientific
048188-1G
048188-500MG
101883-232EA
698.46
USD
101883-232
101883-230
1-Benzyl-3,5-dibromo-4-nitro-1H-pyrazole ≥95%
1-Benzyl-3,5-dibromo-4-nitro-1H-pyrazole
Hazard: Irritant
Formula:
C₁₀H₇Br₂N₃O₂ Melting Pt: 132…133 °C |
MDL Number:
MFCD09972276 CAS Number: 155601-03-9 |
Specification Test Results
Melting point | 132-133°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...