2-Amino-5-bromobenzylalkohol ≥95%

Supplier: Matrix Scientific

Synonyms: (2-Amino-5-bromophenyl)methanol

048182-1G 048182-500MG
101883-206EA 138.46 USD
101883-206 101883-208
2-Amino-5-bromobenzylalkohol ≥95%
2-Amino-5-bromobenzylalkohol

Hazard: Irritant

Formula: C₇H₈BrNO
MW: 202.0500 g/mol
Melting Pt: 114…115 °C
MDL Number: MFCD08234509
CAS Number: 20712-12-3

Order Now

Specification Test Results

Melting point 114-115°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR