5-Bromo-2-morpholinonicotinic acid ≥95%

Supplier: Matrix Scientific

048174-5G 048174-500MG 048174-1G
101883-166EA 2129.23 USD
101883-166 101883-162 101883-164
5-Bromo-2-morpholinonicotinic acid ≥95%
5-Bromo-2-morpholinonicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₁BrN₂O₃
Melting Pt: 168…170 °C
MDL Number: MFCD09972268
CAS Number: 1017783-03-7

Order Now

Specification Test Results

Melting point 168-170°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR