2,4,6-Trichlorophenyl-5-bromo-2-methoxybenzenesulfonate ≥95%
Supplier: Matrix Scientific
048138-5G
048138-500MG
048138-1G
101882-998EA
278.46
USD
101882-998
101883-000
101883-002
2,4,6-Trichlorophenyl-5-bromo-2-methoxybenzenesulfonate ≥95%
2,4,6-Trichlorophenyl-5-bromo-2-methoxybenzenesulfonate
Hazard: Irritant
Formula:
C₁₃H₈BrCl₃O₄S Melting Pt: 112…114 °C |
MDL Number:
MFCD12498733 CAS Number: 1171919-47-3 |
Specification Test Results
Melting point | 112-114°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...