N-(3-Aminophenyl)-N-methylacetamide ≥95%

Supplier: Matrix Scientific

048109-500MG 048109-5G 048109-1G
101882-836EA 280.6 USD
101882-836 101882-840 101882-838
N-(3-Aminophenyl)-N-methylacetamide ≥95%
N-(3-Aminophenyl)-N-methylacetamide

Hazard: Irritant

Formula: C₉H₁₂N₂O
Melting Pt: 116…118 °C
MDL Number: MFCD02657491
CAS Number: 61679-27-4

Order Now

Specification Test Results

Melting point 116-118°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR