2-[4-(2-Hydroxyethyl)piperazino]nicotinonitrile ≥95%

Supplier: Matrix Scientific

048070-500MG 048070-1G
101882-676EA 173.65 USD
101882-676 101882-674
2-[4-(2-Hydroxyethyl)piperazino]nicotinonitrile ≥95%
2-[4-(2-Hydroxyethyl)piperazino]nicotinonitrile

Hazard: Irritant

Formula: C₁₂H₁₆N₄O
Melting Pt: 100…102 °C
MDL Number: MFCD09972255
CAS Number: 1017782-85-2

Order Now

Specification Test Results

Melting point 100-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR