1-(3-Methoxy-4-nitrophenyl)-4-piperidinecarboxylic acid ≥95%
Supplier: Matrix Scientific
048069-1G
048069-5G
048069-500MG
101882-668EA
375.38
USD
101882-668
101882-672
101882-670
1-(3-Methoxy-4-nitrophenyl)-4-piperidinecarboxylic acid ≥95%
1-(3-Methoxy-4-nitrophenyl)-4-piperidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₆N₂O₅ Melting Pt: 217…219 °C |
MDL Number:
MFCD09972254 CAS Number: 1017782-83-0 |
Specification Test Results
Melting point | 217-219°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...