2-Butyl-5,6-dichloro-1H-1,3-benzimidazole ≥95%

Supplier: Matrix Scientific

048065-500MG
101882-648EA 181.54 USD
101882-648
2-Butyl-5,6-dichloro-1H-1,3-benzimidazole ≥95%
2-Butyl-5,6-dichloro-1H-1,3-benzimidazole

Hazard: Irritant

Formula: C₁₁H₁₂Cl₂N₂
MW: 243.14 g/mol
Melting Pt: 149…151 °C
MDL Number: MFCD08064759
CAS Number: 133052-59-2

Order Now

Specification Test Results

Melting point 149-151°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR