2-(4-Phenylpiperazino)-1,3-thiazole-4-carboxylicacid ≥95%
Supplier: Matrix Scientific
048014-1G
048014-500MG
101882-400EA
467.69
USD
101882-400
101882-398
2-(4-Phenylpiperazino)-1,3-thiazole-4-carboxylicacid ≥95%
2-(4-Phenylpiperazino)-1,3-thiazole-4-carboxylicacid
Hazard: Irritant
Formula:
C₁₄H₁₅N₃O₂S MW: 289.36 g/mol Melting Pt: 189…191 °C |
MDL Number:
MFCD09027187 CAS Number: 952182-67-1 |
Specification Test Results
Melting point | 189-191°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...