Methyl-2-methoxyisonicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-methoxyisonicotinate

047996-1G
101882-322EA 44.85 USD
101882-322
Methyl-2-methoxyisonicotinate ≥95%
Methyl 2-methoxyisonicotinate

Hazard: Irritant

Formula: C₈H₉NO₃
MW: 167.1620 g/mol
Melting Pt: 36…38 °C
MDL Number: MFCD08436071
CAS Number: 26156-51-4

Order Now

Specification Test Results

Melting point 36-38°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR