2-Ethyl-1-methyl-5-nitro-1H-1,3-benzimidazole ≥95%

Supplier: Matrix Scientific

047992-500MG
101882-308EA 375.38 USD
101882-308
2-Ethyl-1-methyl-5-nitro-1H-1,3-benzimidazole ≥95%
2-Ethyl-1-methyl-5-nitro-1H-1,3-benzimidazole

Hazard: Irritant

Formula: C₁₀H₁₁N₃O₂
Melting Pt: 192…194 °C
MDL Number: MFCD08688844
CAS Number: 286837-93-2

Order Now

Specification Test Results

Melting point 192-194°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR