1-Methyl-5-nitro-1H-1,3-benzimidazole ≥95%

Supplier: Matrix Scientific

047989-500MG
101882-298EA 266.8 USD
101882-298
1-Methyl-5-nitro-1H-1,3-benzimidazole ≥95%
1-Methyl-5-nitro-1H-1,3-benzimidazole

Hazard: Irritant

Formula: C₈H₇N₃O₂
Melting Pt: 210…212 °C
MDL Number: MFCD00628958
CAS Number: 5381-78-2

Order Now

Specification Test Results

Melting point 210-212°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR