2-Oxo-6-(2-pyridinyl)-1,2-dihydro-3-pyridine carboxylic acid ≥95%
Supplier: Matrix Scientific
047983-500MG
047983-5G
047983-1G
101882-270EA
375.38
USD
101882-270
101882-274
101882-272
2-Oxo-6-(2-pyridinyl)-1,2-dihydro-3-pyridine carboxylic acid ≥95%
2-Oxo-6-(2-pyridinyl)-1,2-dihydro-3-pyridine carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₈N₂O₃ MW: 216.2 g/mol Melting Pt: 304…306 °C |
MDL Number:
MFCD11841017 CAS Number: 56304-42-8 |
Specification Test Results
Melting point | 304-306°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...