tert-Butyl-4-[(4-cyano-2-pyridinyl)oxy]tetrahydro-1(2H)-pyridinecarboxylate ≥95%
Supplier: Matrix Scientific
047973-500MG
047973-1G
101882-236EA
358.46
USD
101882-236
101882-234
tert-Butyl-4-[(4-cyano-2-pyridinyl)oxy]tetrahydro-1(2H)-pyridinecarboxylate ≥95%
tert-Butyl-4-[(4-cyano-2-pyridinyl)oxy]tetrahydro-1(2H)-pyridinecarboxylate
Hazard: Irritant
Formula:
C₁₆H₂₁N₃O₃ MW: 303.36 g/mol Melting Pt: 123…124 °C |
MDL Number:
MFCD09877713 CAS Number: 1065484-24-3 |
Specification Test Results
Melting point | 123-124°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...