2-Piperazino-4-pyrimidinecarbonitrile ≥95%
Supplier: Matrix Scientific
047970-1G
047970-500MG
101882-218EA
733.85
USD
101882-218
101882-220
2-Piperazino-4-pyrimidinecarbonitrile ≥95%
2-Piperazino-4-pyrimidinecarbonitrile
Hazard: Irritant
Formula:
C₉H₁₁N₅ Melting Pt: 104…105 °C |
MDL Number:
MFCD11841014 CAS Number: 1135283-54-3 |
Specification Test Results
Melting point | 104-105°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...