Tetrahydro-1H-pyrrolo[1,2-c]imidazole-1,3(2H)-dione ≥95%

Supplier: Matrix Scientific

047964-1G 047964-500MG
101882-192EA 461.54 USD
101882-192 101882-190
Tetrahydro-1H-pyrrolo[1,2-c]imidazole-1,3(2H)-dione ≥95%
Tetrahydro-1H-pyrrolo[1,2-c]imidazole-1,3(2H)-dione

Hazard: Irritant

Formula: C₆H₈N₂O₂
Melting Pt: 160…162 °C
MDL Number: MFCD01851016
CAS Number: 5768-79-6

Order Now

Specification Test Results

Melting point 160-162°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR