3-[(Dimethylamino)sulfonyl]benzenecarboxylic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
3-(N,N-Dimethylsulphamoyl)benzoic acid
047935-1G
047935-500MG
047935-5G
101882-062EA
49.23
USD
101882-062
101882-066
101882-064
3-[(Dimethylamino)sulfonyl]benzenecarboxylic acid ≥97%
3-[(Dimethylamino)sulfonyl]benzenecarboxylic acid
Hazard: Irritant
Formula:
C₉H₁₁NO₄S MW: 229.26 g/mol Melting Pt: 174…176 °C |
MDL Number:
MFCD02708231 CAS Number: 7326-73-0 |
Specification Test Results
Melting point | 174-176°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...