2-(4-Methoxyanilino)-2-(2-thienyl)acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: [(4-Methoxyphenyl)amino](thien-2-yl)acetic acid

047931-500MG
101882-044EA 747.69 USD
101882-044
2-(4-Methoxyanilino)-2-(2-thienyl)acetic acid ≥95%
2-(4-Methoxyanilino)-2-(2-thienyl)acetic acid

Hazard: Irritant

Formula: C₁₃H₁₃NO₃S
MW: 263.32 g/mol
Melting Pt: 168…170 °C
MDL Number: MFCD04116995
CAS Number: 199461-24-0

Order Now

Specification Test Results

Melting point 168-170°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR