tert-Butyl-4-[5-bromo-3-(methoxycarbonyl)-2-pyridinyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 5-bromo-2-piperazin-1-ylpyridine-3-carboxylic acid, N4-BOC protected
047930-5G
047930-500MG
047930-1G
101882-040EA
378.46
USD
101882-040
101882-038
101882-042
tert-Butyl-4-[5-bromo-3-(methoxycarbonyl)-2-pyridinyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-[5-bromo-3-(methoxycarbonyl)-2-pyridinyl]tetrahydro-1(2H)-pyrazinecarboxylate
Hazard: Irritant
Formula:
C₁₆H₂₂BrN₃O₄ MW: 400.28 g/mol Melting Pt: 122…124 °C |
MDL Number:
MFCD09864832 CAS Number: 1000018-22-3 |
Specification Test Results
Melting point | 122-124°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...