4-(4-Bromophenyl)-5-(methylsulfonyl)-1,3-thiazol-2-amine ≥95%

Supplier: Matrix Scientific

047928-500MG
101882-036EA 921.54 USD
101882-036
4-(4-Bromophenyl)-5-(methylsulfonyl)-1,3-thiazol-2-amine ≥95%
4-(4-Bromophenyl)-5-(methylsulfonyl)-1,3-thiazol-2-amine

Hazard: Irritant

Formula: C₁₀H₉BrN₂O₂S₂
Melting Pt: 253…255 °C
MDL Number: MFCD09972236
CAS Number: 1017782-62-5

Order Now

Specification Test Results

Melting point 253-255°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR