3,5-Dibromo-1H-indole-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
047922-500MG
047922-1G
101882-018EA
467.69
USD
101882-018
101882-016
3,5-Dibromo-1H-indole-2-carboxylic acid ≥95%
3,5-Dibromo-1H-indole-2-carboxylic acid
Hazard: Irritant
Formula:
C₉H₅Br₂NO₂ Melting Pt: 230 °C (dec.) |
MDL Number:
MFCD09027182 CAS Number: 98591-49-2 |
Specification Test Results
Melting point | 230°C(dec) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...