Methyl-3-[(4-bromobenzoyl)(methyl)amino]-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

047906-5G 047906-500MG 047906-1G
101881-932EA 278.46 USD
101881-932 101881-934 101881-936
Methyl-3-[(4-bromobenzoyl)(methyl)amino]-2-thiophenecarboxylate ≥95%
Methyl-3-[(4-bromobenzoyl)(methyl)amino]-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₂BrNO₃S
Melting Pt: 114…116 °C
MDL Number: MFCD10758101
CAS Number: 1033463-43-2

Order Now

Specification Test Results

Melting point 114-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR