tert-Butyl-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}tetrahydro-1(2H)-pyridinecarboxylate ≥95%
Supplier: Matrix Scientific
047847-500MG
047847-1G
101881-692EA
698.46
USD
101881-692
101881-690
tert-Butyl-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}tetrahydro-1(2H)-pyridinecarboxylate ≥95%
tert-Butyl-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}tetrahydro-1(2H)-pyridinecarboxylate
Hazard: Irritant
Formula:
C₁₆H₂₁ClF₃N₃O₂ MW: 379.81 g/mol Melting Pt: 95…96 °C |
MDL Number:
MFCD11553061 CAS Number: 1092352-86-7 |
Specification Test Results
Melting point | 95-96°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...