2-Chloro-N'-hydroxy-4-pyridinecarboximidamide ≥95%

Supplier: Matrix Scientific

047844-500MG 047844-1G
101881-680EA 467.69 USD
101881-680 101881-678
2-Chloro-N'-hydroxy-4-pyridinecarboximidamide ≥95%
2-Chloro-N'-hydroxy-4-pyridinecarboximidamide

Hazard: Irritant

Formula: C₆H₆ClN₃O
Melting Pt: 162…164 °C
MDL Number: MFCD11052368
CAS Number: 857653-85-1

Order Now

Specification Test Results

Melting point 162-164°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR