7-Bromo-2-(chloromethyl)-5-methoxy-1,3-benzoxazole ≥95%
Supplier: Matrix Scientific
047843-1G
047843-500MG
101881-674EA
747.69
USD
101881-674
101881-676
7-Bromo-2-(chloromethyl)-5-methoxy-1,3-benzoxazole ≥95%
7-Bromo-2-(chloromethyl)-5-methoxy-1,3-benzoxazole
Hazard: Irritant
Formula:
C₉H₇BrClNO₂ Melting Pt: 107…109 °C |
MDL Number:
MFCD11553060 CAS Number: 1092352-84-5 |
Specification Test Results
Melting point | 107-109°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...