N'-Hydroxy-2,2-dimethylpropanimidamide ≥95%
Supplier: Matrix Scientific
Synonyms:
(1Z)-N'-hydroxy-2,2-dimethylpropanimidamide
, Propanimidamide, N'-hydroxy-2,2-dimethyl-
047835-5G
047835-1G
047835-500MG
101881-644EA
172.31
USD
101881-644
101881-646
101881-648
N'-Hydroxy-2,2-dimethylpropanimidamide ≥95%
N'-Hydroxy-2,2-dimethylpropanimidamide
Hazard: Irritant
Formula:
C₅H₁₂N₂O MW: 116.16 g/mol Melting Pt: 113…115 °C |
MDL Number:
MFCD03426250 CAS Number: 42956-75-2 |
Specification Test Results
Melting point | 113-115°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...