2,4-Dimethyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylic acid ≥95%
Supplier: Matrix Scientific
047822-1G
047822-500MG
101881-596EA
1109.23
USD
101881-596
101881-598
2,4-Dimethyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylic acid ≥95%
2,4-Dimethyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₁₁NO₄ Melting Pt: 210…212 °C |
MDL Number:
MFCD11553054 CAS Number: 1092352-74-3 |
Specification Test Results
Melting point | 210-212°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...