1-Benzyl-4-nitro-1H-pyrazole ≥95%

Supplier: Matrix Scientific

047808-1G 047808-500MG
101881-534EA 280.6 USD
101881-534 101881-536
1-Benzyl-4-nitro-1H-pyrazole ≥95%
1-Benzyl-4-nitro-1H-pyrazole

Hazard: Irritant

Formula: C₁₀H₉N₃O₂
Melting Pt: 56…58 °C
MDL Number: MFCD03087886
CAS Number: 88095-61-8

Order Now

Specification Test Results

Melting point 56-58°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR