N-Methyl-1,3-benzothiazol-2-amine ≥95%

Supplier: Matrix Scientific

047802-500MG 047802-1G 047802-5G
101881-510EA 262.2 USD
101881-510 101881-512 101881-508
N-Methyl-1,3-benzothiazol-2-amine ≥95%
N-Methyl-1,3-benzothiazol-2-amine

Hazard: Irritant

Formula: C₈H₈N₂S
Melting Pt: 140…142 °C
MDL Number: MFCD00035877
CAS Number: 16954-69-1

Order Now

Specification Test Results

Melting point 140-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR