N-Methyl-1,3-benzoxazol-2-amine ≥95%
Supplier: Matrix Scientific
Synonyms:
N-Methylbenzo[d]oxazol-2-amine
047801-5G
047801-500MG
047801-1G
101881-502EA
138.46
USD
101881-502
101881-504
101881-506
N-Methyl-1,3-benzoxazol-2-amine ≥95%
N-Methyl-1,3-benzoxazol-2-amine
Hazard: Irritant
Formula:
C₈H₈N₂O MW: 148.17 g/mol Melting Pt: 75…77 °C |
MDL Number:
MFCD00613415 CAS Number: 19776-98-8 |
Specification Test Results
Melting point | 75-77°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...