5-Bromo-1-methyl-1H-imidazole-2-carbaldehyde ≥95%

Supplier: Matrix Scientific

047797-500MG
101881-484EA 558.9 USD
101881-484
5-Bromo-1-methyl-1H-imidazole-2-carbaldehyde ≥95%
5-Bromo-1-methyl-1H-imidazole-2-carbaldehyde

Hazard: Irritant

Formula: C₅H₅BrN₂O
MW: 189.02 g/mol
Melting Pt: 70…73 °C
MDL Number: MFCD09027273
CAS Number: 79326-88-8

Order Now

Specification Test Results

Melting point 70-73°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR