Methyl-4-oxo-4,5,6,7-tetrahydro-1-benzofuran-3-carboxylate ≥95%

Supplier: Matrix Scientific

047769-1G 047769-500MG 047769-5G
101881-340EA 345 USD
101881-340 101881-342 101881-338
Methyl-4-oxo-4,5,6,7-tetrahydro-1-benzofuran-3-carboxylate ≥95%
Methyl-4-oxo-4,5,6,7-tetrahydro-1-benzofuran-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Melting Pt: 104…106 °C
MDL Number: MFCD09027158
CAS Number: 82584-78-9

Order Now

Specification Test Results

Melting point 104-106°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR