Methyl-4-(2-piperazinoethoxy)benzenecarboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

047766-500MG 047766-1G 047766-5G
101881-322EA 375.38 USD
101881-322 101881-324 101881-326
Methyl-4-(2-piperazinoethoxy)benzenecarboxylate hydrochloride ≥95%
Methyl-4-(2-piperazinoethoxy)benzenecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁ClN₂O₃
Melting Pt: >230 °C (dec.)
MDL Number: MFCD09027156
CAS Number: 1170473-34-3

Order Now

Specification Test Results

Melting point 230°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR