2,6-Difluorobenzenecarbaldehyde oxime ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Difluorobenzaldoxime

047761-1G 047761-5G 047761-500MG
101881-296EA 135.7 USD
101881-296 101881-298 101881-300
2,6-Difluorobenzenecarbaldehyde oxime ≥95%
2,6-Difluorobenzenecarbaldehyde oxime

Hazard: Irritant

Formula: C₇H₅F₂NO
MW: 157.12 g/mol
Melting Pt: 114…116 °C
MDL Number: MFCD01764489
CAS Number: 19064-16-5

Order Now

Specification Test Results

Melting point 114-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR