2-(4-Fluorophenyl)-1,3-thiazolane ≥95%
Supplier: Matrix Scientific
047760-1G
047760-500MG
047760-5G
101881-290EA
181.54
USD
101881-290
101881-292
101881-294
2-(4-Fluorophenyl)-1,3-thiazolane ≥95%
2-(4-Fluorophenyl)-1,3-thiazolane
Hazard: Irritant
Formula:
C₉H₁₀NFS Melting Pt: 80…82 °C |
MDL Number:
MFCD01681205 CAS Number: 67086-80-0 |
Specification Test Results
Melting point | 80-82°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...