Ethyl-2-(hydroxyimino)-2-(1H-imidazol-1-yl)acetate ≥95%
Supplier: Matrix Scientific
047751-1G
047751-5G
047751-500MG
101881-252EA
806.15
USD
101881-252
101881-248
101881-250
Ethyl-2-(hydroxyimino)-2-(1H-imidazol-1-yl)acetate ≥95%
Ethyl-2-(hydroxyimino)-2-(1H-imidazol-1-yl)acetate
Hazard: Irritant
Formula:
C₇H₉N₃O₃ MW: 183.17 g/mol Melting Pt: 142…144 °C |
MDL Number:
MFCD09027146 CAS Number: 95080-92-5 |
Specification Test Results
Melting point | 142-144°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...