2-(4-Ethoxyphenyl)-1,3-thiazolane ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(4-Ethoxyphenyl)-1,3-thiazolidine
047749-5G
047749-1G
047749-500MG
101881-240EA
532.31
USD
101881-240
101881-236
101881-238
2-(4-Ethoxyphenyl)-1,3-thiazolane ≥95%
2-(4-Ethoxyphenyl)-1,3-thiazolane
Hazard: Irritant
Formula:
C₁₁H₁₅NOS Melting Pt: 96…98 °C |
MDL Number:
MFCD09027144 CAS Number: 937602-47-6 |
Specification Test Results
Melting point | 96-98°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...