2-[3-Fluoro-4-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylic acid ≥95%
Supplier: Matrix Scientific
047739-500MG
047739-1G
047739-5G
101881-188EA
747.69
USD
101881-188
101881-190
101881-192
2-[3-Fluoro-4-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylic acid ≥95%
2-[3-Fluoro-4-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₅F₄NO₂S Melting Pt: 178…180 °C |
MDL Number:
MFCD09027138 CAS Number: 937602-43-2 |
Specification Test Results
Melting point | 178-180°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...