3-(1-Pyrrolidinylsulfonyl)benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Pyrrolidin-1-ylsulphonyl)benzoic acid

047737-500MG 047737-1G
101881-180EA 107.69 USD
101881-180 101881-182
3-(1-Pyrrolidinylsulfonyl)benzenecarboxylic acid ≥95%
3-(1-Pyrrolidinylsulfonyl)benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₄S
MW: 255.29 g/mol
Melting Pt: 149…151 °C
MDL Number: MFCD00814149
CAS Number: 7326-79-6

Order Now

Specification Test Results

Melting point 149-151°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR