Ethyl-2-[3-fluoro-4-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

047733-500MG 047733-1G 047733-5G
101881-156EA 556.92 USD
101881-156 101881-158 101881-160
Ethyl-2-[3-fluoro-4-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylate ≥95%
Ethyl-2-[3-fluoro-4-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₃H₉F₄NO₂S
MW: 319.28 g/mol
Melting Pt: 111…113 °C
MDL Number: MFCD09027134
CAS Number: 937602-41-0

Order Now

Specification Test Results

Melting point 111-113°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR