5-Chloro-3-ethynyl-2-pyridinamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-chloro-3-ethynylpyridine

046632-500MG
101876-346EA 556.92 USD
101876-346
5-Chloro-3-ethynyl-2-pyridinamine ≥95%
5-Chloro-3-ethynyl-2-pyridinamine

Hazard: Irritant

Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Melting Pt: 118…119 °C
MDL Number: MFCD10758079
CAS Number: 866318-88-9

Order Now

Specification Test Results

Melting point 118-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR