Methyl-4-(cyclohexylamino)-3-nitrobenzenecarboxylate ≥95%
Supplier: Matrix Scientific
046611-500MG
046611-5G
046611-1G
101876-270EA
806.15
USD
101876-270
101876-266
101876-268
Methyl-4-(cyclohexylamino)-3-nitrobenzenecarboxylate ≥95%
Methyl-4-(cyclohexylamino)-3-nitrobenzenecarboxylate
Hazard: Irritant
Formula:
C₁₄H₁₈N₂O₄ Melting Pt: 132…134 °C |
MDL Number:
MFCD03855459 CAS Number: 503859-26-5 |
Specification Test Results
Melting point | 132-134°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...