(4'-Nitro[1,1'-biphenyl]-2-yl)methanol ≥95%

Supplier: Matrix Scientific

046609-500MG
101876-258EA 921.54 USD
101876-258
(4'-Nitro[1,1'-biphenyl]-2-yl)methanol ≥95%
(4'-Nitro[1,1'-biphenyl]-2-yl)methanol

Hazard: Irritant

Formula: C₁₃H₁₁NO₃
Melting Pt: 119…121 °C
MDL Number: MFCD10758069
CAS Number: 500294-19-9

Order Now

Specification Test Results

Melting point 119-121°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR