(3'-Nitro[1,1'-biphenyl]-4-yl)methanol ≥95%

Supplier: Matrix Scientific

046608-500MG
101876-256EA 921.54 USD
101876-256
(3'-Nitro[1,1'-biphenyl]-4-yl)methanol ≥95%
(3'-Nitro[1,1'-biphenyl]-4-yl)methanol

Hazard: Irritant

Formula: C₁₃H₁₁NO₃
Melting Pt: 84…86 °C
MDL Number: MFCD06803033
CAS Number: 880158-11-2

Order Now

Specification Test Results

Melting point 84-86°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR