4-Bromo-1H-indazole ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromoindazole

046607-5G
101876-254EA 36.92 USD
101876-254
4-Bromo-1H-indazole ≥95%
4-Bromo-1H-indazole

Hazard: Irritant

Formula: C₇H₅BrN₂
MW: 197.04 g/mol
Melting Pt: 165…167 °C
MDL Number: MFCD05664001
CAS Number: 186407-74-9

Order Now

Specification Test Results

Melting point 165-167°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR