5-Nitro-1-benzofuran-2-carbaldehyde ≥95%
Supplier: Matrix Scientific
046606-500MG
046606-1G
101876-250EA
807.69
USD
101876-250
101876-248
5-Nitro-1-benzofuran-2-carbaldehyde ≥95%
5-Nitro-1-benzofuran-2-carbaldehyde
Hazard: Irritant
Formula:
C₉H₅NO₄ Melting Pt: 161…163 °C |
MDL Number:
MFCD06255199 CAS Number: 23145-18-8 |
Specification Test Results
Melting point | 161-163°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...